Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chloride
Sodium chloride 9% (w/v) in aqueous solution
Sodium chloride 9% (w/v) in aqueous solution
Sodium chloride 9% (w/v) in aqueous solution
DIUKFE1322
:
DISTRIBUTOR
:
Sodium chloride 9% (w/v) in aqueous solution
DIUKFE1322
:
DISTRIBUTOR
:
Pack type:
Viaflo
Pk:
30
Seq:
0030
Pk Info:
30 x 250 mL
:
:
58.44 g/mol
:
Ambient
:
231-598-3
:
7647-14-5
:
NaCl
:
MFCD00003477