Phenol liquified ≥89.0%, Bioreagent
Catalog # E357-500ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Distilling Range:≥182.5 °C
- Nonvolatile Residue:≥0.05%
- Purity (%):89.0
Specification Test Results
About this item
:
:94.11 g/mol
Melting Pt:40°C-42°C
Storage Temperature:Ambient
EINECS:203-632-7
UN:2821
ADR:6.1,II
:GHS06
:GHS05
:GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143