Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenol liquified
Phenol liquified ≥89%, Sigma-Aldrich®
Phenol liquified ≥89%, Sigma-Aldrich®
:
Merck
Phenol liquified ≥89%, Sigma-Aldrich®
:
Merck
:
108-95-2
:
:
C₆H₅OH
:
MFCD00002143
:
94.11 g/mol
:
6.1,II
:
M-Clarity™ quality level = MQ200
:
203-632-7
:
2821
:
GHS06
:
GHS05
:
GHS08
:
Ambient