Rifampicin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13292-46-1
:
Formula:C₄₃H₅₈N₄O₁₂
MDL Number:MFCD00151389
:822.95 g/mol
Merck Index:12,08382
Melting Pt:183…188 °C
:M-Clarity™ quality level = MQ200
EINECS:236-312-0