Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rifampicin
Rifampicin, Millipore®
Rifampicin, Millipore®
Rifampicin, Millipore®
Supplier:
Merck Millipore (Calbiochem)
Certificates
Expand image
Rifampicin, Millipore®
Supplier:
Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:
13292-46-1
:
Formula:
C₄₃H₅₈N₄O₁₂
MDL Number:
MFCD00151389
:
822.95 g/mol
Merck Index:
12,08382
Melting Pt:
183…188 °C
EINECS:
236-312-0
Related Information
Documentation
Certificates
Search by Lot Number
Documentation