Order Entry
Germany
ContactUsLinkComponent
Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) Reag. Ph. Eur. for the analysis of sugar, Fluka™
Corrosive
Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) Reag. Ph. Eur. for the analysis of sugar, Fluka™
Certificates
Corrosive
Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) Reag. Ph. Eur. for the analysis of sugar, Fluka™

Specification Test Results

  • Density (D 20/4):
    1.23 - 1.25
  • Application test:
    complying

Specification Test Results

Frequently Bought Together