Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
G418 disulphate
G418 disulphate, Sigma-Aldrich®
G418 disulphate, Sigma-Aldrich®
G418 disulphate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
G418 disulphate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
108321-42-2
:
Formula:
C₂₀H₄₄N₄O₁₈S₂
MDL Number:
MFCD00058314
:
692.72 g/mol
:
M-Clarity™ quality level = MQ200
:
GHS07
Storage Temperature:
Ambient