cis-Platin
Catalog # MOLEM68655191
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:300.05 g/mol
Merck Index:13,02341
Density:3.738 g/cm³ (20 °C)
EINECS:239-733-8
UN:3288
ADR:6.1,II
CAS Number:15663-27-1
Formula:H₆Cl₂N₂Pt
MDL Number:MFCD00011623