L(+)-Rhamnose monohydrate, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:10030-85-0
:
Formula:C₆H₁₂O₅·1H₂O
MDL Number:MFCD00149363
:182.17 g/mol
Merck Index:12,08338
Melting Pt:85…95 °C
Density:1.47 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:222-793-4