Order Entry
Germany
ContactUsLinkComponent
L(+)-Rhamnose monohydrate
L(+)-Rhamnose monohydrate
Catalog # MOLEM71239713
Supplier:  Molekula
CAS Number:  
L(+)-Rhamnose monohydrate
Catalog # MOLEM71239713
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :182.17 g/mol
Merck Index:12,08338
Melting Pt:85…95 °C
Density:1.47 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:222-793-4
CAS Number:10030-85-0
Formula:C₆H₁₂O₅·1H₂O
MDL Number:MFCD00149363