Lactose monohydrate
Glass bottle for solids, 250 g
Supplier: CAESAR LORENTZ
Lactose monohydrate
Catalog # CSLO2582.0250
Supplier: CAESAR LORENTZ
Marketsource
CAS Number: 10039-26-6
Jump to:
Product Details & Documents
Synonyms: Milk sugar, Lactobiose
- CAS Number:10039-26-6
- Formula:C₁₂H₂₂O₁₁·1H₂O
- Molecular Weight:360.32 g/mol
- Melting Pt:223 °C
- Storage Temperature:Ambient
- EINECS:200-559-2
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



