Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ethanol-[D₆]

Glass bottle, 1 mL
Catalog # LBCLC2D6O1
Flammable
Marketsource
Supplier:   LABC LABORTECHNIK
Ethanol-[D₆]
Catalog # LBCLC2D6O1
Supplier:  LABC LABORTECHNIK
Marketsource
CAS Number:  1516-08-1
Ethanol-[D₆]

Glass bottle, 1 mL

CAS Number:  1516-08-1
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  Methyl carbinol-D6, Hydroxy ethane-D6, Ethanol-[²H₆], Ethyl alcohol-D6
  • CAS Number:1516-08-1
  • Formula:D₃CCD₂OD
  • MDL Number:MFCD00051020
  • Molecular Weight:52.02 g/mol
  • Boiling Pt:78 °C (1013 hPa)
  • Melting Pt:–114,5 °C
  • Melting Pt:–114.5 °C
  • Density:0,801 g/cm³ (20 °C)
  • Flash Pt:13 °C
  • EINECS:216-162-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications