Vanadyl sulphate pentahydrate, TECHNICAL
Catalog # 28900.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:253.08 g/mol
Density:2 g/cm³ (20 °C)
EINECS:248-652-7
UN:2931
ADR:6.1,II
:GHS07
CAS Number:12439-96-2
Formula:VOSO₄·5H₂O
MDL Number:MFCD00149933