Order Entry
Germany
ContactUsLinkComponent
Sodium bisulphite 50% (w/v) in aqueous solution, TECHNICAL
Irritant
Sodium bisulphite 50% (w/v) in aqueous solution, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Sodium bisulphite 50% (w/v) in aqueous solution, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (W/V):
    Min. 50 %
  • Ionised chlorine:
    Max. 0.05 %

Specification Test Results