Order Entry
Germany
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) ≥98% analytical reagent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) ≥98% analytical reagent
Catalog # 27963.101
Supplier:  VWR Chemicals
CAS Number:  
undefined
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) ≥98% analytical reagent
Catalog # 27963.101
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • 4-Nitrophenol:
    Max. 0.2 %
  • Water:
    Max. 30 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

About this item

 :
Melting Pt:> 300 °C
 :371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
CAS Number:333338-18-4
 :4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288