Order Entry
Germany
Orders LinkContactUsLinkComponent
Nuclear Fast Red, TECHNICAL

Nuclear Fast Red, TECHNICAL

Glass bottle, 5 g
Catalog # 27416.101
Supplier:   VWR Chemicals
Nuclear Fast Red, TECHNICAL
Catalog # 27416.101
Supplier:  VWR Chemicals
CAS Number:  6409-77-4
Nuclear Fast Red, TECHNICAL
Nuclear Fast Red, TECHNICAL

Glass bottle, 5 g

CAS Number:  6409-77-4
Price
219,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
  • CAS Number:6409-77-4
  • Formula:C₁₄H₈NNaO₇S
  • MDL Number:MFCD00078493
  • Molecular Weight:357.28 g/mol
  • Merck Index:13,06512
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:229-088-0
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications