1-Naphtholbenzeine analytical reagent
Catalog # 25768.136
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Suited for acid-base indicator:Passes test
Specification Test Results
About this item
:
:374.44 g/mol
Melting Pt:243…245 °C
EINECS:205-656-3
:GHS07
Storage Temperature:Ambient
CAS Number:145-50-6
:6948-88-5
Formula:C₂₇H₁₈O₂
MDL Number:MFCD00078492