Order Entry
Germany
ContactUsLinkComponent
Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity
Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity
Catalog # 22525.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity
Catalog # 22525.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Viscosity (25°C; 2 %; water):
    300 - 600 mPa.s
  • Water:
    Max. 8 %

Specification Test Results

About this item

 :
Melting Pt:>300 °C
Storage Temperature:Ambient
CAS Number:9004-32-4
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472