Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Apiezon® Wax W
Apiezon® wax W 40
Apiezon® wax W 40
Apiezon® wax W 40
Catalog # 96030.340
Supplier:
M I MATERIALS
CAS Number:
Expand image
Apiezon® wax W 40
Catalog # 96030.340
Supplier:
M I MATERIALS
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
250 g
Seq:
3545
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
Product Details