ContactUsLinkComponent
VWR® Boiling Stones ('Anti-Bumping'), Granules
VWR® Boiling Stones ('Anti-Bumping'), Granules
Catalog # 330093Y
Supplier:  VWR Chemicals
undefined
VWR® Boiling Stones ('Anti-Bumping'), Granules
Catalog # 330093Y
Supplier:  VWR Chemicals
Specifications
  • Description:
    Anti-bumping granules
  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
Specifications
About this item
Granules of fused alumina, to reduce 'bumping' in boiling liquids.
  • Particle size (1-4 mm): min.96%