Order Entry
Germany
ContactUsLinkComponent
Silica monitor reagent (sodium molybdate) for seres FG 504B silicameter
Silica monitor reagent (sodium molybdate) for seres FG 504B silicameter
Catalog # 161277V
Supplier:  VWR Chemicals
CAS Number:  
undefined
Silica monitor reagent (sodium molybdate) for seres FG 504B silicameter
Catalog # 161277V
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :205.92 g/mol
EINECS:231-551-7
CAS Number:7631-95-0
Formula:Na₂MoO₄