Order Entry
Germany
ContactUsLinkComponent
VWR® Weighing Paper, Grade 531
VWR® Weighing Paper, Grade 531
Catalog # 516-0318
Supplier:  VWR Collection
undefined
VWR® Weighing Paper, Grade 531
Catalog # 516-0318
Supplier:  VWR Collection

Specifications

  • Size:
    100×100 mm
  • Thickness:
    0,06 mm
  • Pk:
    250
  • Weight:
    45 g/m²

Specifications

About this item

For weighing a range of substances (viscous products, powders, paste and liquids).