ContactUsLinkComponent
Spatulas, Trulla type
Spatulas, Trulla type
  231-0393
undefined
Spatulas, Trulla type
  231-0393
  • Length:
    175 mm
  • Pk:
    5
Stainless steel 18/8 non-magnetic, bright finish. Double ended spatulas with trowel shaped scoops for handling large quantities of material.
  • Excellent thermal resistance, autoclavable
  • Excellent chemical resistance
  • Quick and easy to clean