Order Entry
Germany
ContactUsLinkComponent
VWR®, Smoke Point standard
VWR®, Smoke Point standard
Catalog # 99125.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Smoke Point standard
Catalog # 99125.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Smoke Point standard jet aviation fuel, 24.47 mm
  • Test method:
    ASTM D1322 / IP 57

Specifications

About this item

Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Smoke Point ASTM D1322 / IP 57.

  • Certified in accordance with ASTM / EN or IP test method protocols
  • Fully traceable to international standards
  • Tamper-evident security packaging
Certifications: Fully certified in accordance with ISO 17034 under accreditation by UKAS.