Order Entry
Germany
ContactUsLinkComponent
trans-4-tert-Butylcyclohexanol ≥95%
Irritant
trans-4-tert-Butylcyclohexanol ≥95%
Catalog # ABCRTCB1271
Supplier:  abcr
CAS Number:  
Irritant
trans-4-tert-Butylcyclohexanol ≥95%
Catalog # ABCRTCB1271
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 g

Specifications

About this item

 :
 :156.27 g/mol
Melting Pt:80°C
 :GHS07
CAS Number:21862-63-5
Formula:C₁₀H₂₀O
MDL Number:MFCD00064952