Order Entry
Germany
ContactUsLinkComponent
Human Recombinant GALECTIN-1 (Animal free) (from E. coli)
Human Recombinant GALECTIN-1 (Animal free) (from E. coli)
Certificates
undefined
Human Recombinant GALECTIN-1 (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P09382
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    S-LAC lectin-I|L-14|Beta-galactoside-binding lectin L-14-I|galaptin
  • UniProtKB:
    P09382
  • Protein/peptide name:
    Galectin-1
  • Purity:
    ≥95%
  • Molecular weight:
    14,7 kDa
  • Sequence:
    MACGLVASNL NLKPGECLRV RGEVAPDAKS FVLNLGKDSN NLCLHFNPRF NAHGDANTIV CNSKDGGAWG TEQREAVFPF QPGSVAEVCI TFDQANLTVK LPDGYEFKFP NRLNLEAINY MAADGDFKIK CVAFD
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together