Specifications
- Colour:Beige
- Description:Hi-Strength 90 spray adhesive
- Pk:12
Specifications
About this item
3M™ hi-strength 90 spray adhesive is an extremely versatile, fast-drying spray adhesive that bonds strongly to a wide range of materials. A permanent bond ensures fast results to keep projects moving ahead.
- Provides professional, industrial strength adhesion for demanding applications
- Creates soft, non-dimpling adhesive lines for a clean appearance
- High temperature and moisture resistance
- Variable-width spray pattern for more control and minimum waste
The spray formula is easy to dispense, offering a variable-width spray pattern, and applies directly onto surfaces and materials where needed. This spray adhesive offers a high initial grab, sufficient bonding range, and a fast dry time. The high solids content provides higher per square foot coverage than competing spray adhesives. The combination of maximum strength bond and clean appearance make it ideal for projects of all kinds.
Recommended applications include general maintenance and repair projects, attaching fiberglass insulation and drywall corner beads, bonding thin films, foils and fabrics, picture framing and mounting displays, crafts and hobbies, and installing acoustic panels.