Safety stripe tape, 5702
Catalog # MMMA5702
Supplier: 3M
Specifications
- Colour:Yellow/black
- Description:Safety stripe tape, rubber adhesive, vinyl backing, 50 mm width, 33 m length
- Pk:1 Roll
Specifications
About this item
This tape is ideal for marking lanes, hazardous areas, protruding equipment, and low hanging objects in factories, warehouses, and hospitals. Vivid, high-visibility yellow and black diagonal stripes that lock into the vinyl alert people about hazardous conditions.
- Applies instantly and is ready to use immediately
- Yellow and black stripes are locked into the vinyl for permanent high visibility
- Stretches and conforms to curved surfaces and around corners for a tight fit
- Resists abrasion, scuffing, moisture, and weathering for long service life
- Used to identify and mark traffic areas and physical hazards
Unlike non-adhesive caution or barricade tape, this tape features a rubber adhesive that sticks quickly to a variety of surfaces, and removes just as easily, leaving no messy residue. Using vinyl tape for lane marking instead of painting eliminates the need to ventilate paint solvents. And since vinyl tapes can be removed quickly and cleanly, they are also a faster, more versatile, and less costly option than painting.
Adhesion strength - range of 21 N/100 mm.