VWR®, Dot Printer for VWR® Balances
611-4790
- Description:TX-110 dot printer for balances
- Seq:001
- Pk:1
The TX-110 provides high speed dot printing and features a 184×64 OLED display.
- Printouts are suitable for long-term preservation
- ISO/GLP compliant data printout
- Built-in real-time clock
- Sample name and batch number can be input and printed
- Sum, mean and formulation function