Acetonitrile, dried, Biotechnology Grade (max. 10 ppm H₂O), Burdick & Jackson™
Catalog # HONC65739-7L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Stainless steel can
- Pk:7 L
- Seq:5202
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Non-volatile matter:effective %
- Water (Karl Fischer):Max. 0.001 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0001 %
Specification Test Results
About this item
:
:41.05 g/mol
Boiling Pt:81.6 °C
Density:0.7857 g/cm3
Flash Pt:6 °C
UN:1648
ADR:3,II
:GHS07
:GHS02
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878