Tetrahydrofuran, Burdick & Jackson™
Catalog # HONC65529-177KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:177 kg
- Seq:4660
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.05 %
- Peroxides (as H₂O₂):Max. 0.005 %
- Assay BHT (GC):100 - 300 ppm
Specification Test Results
About this item
:
:72.11 g/mol
Boiling Pt:65 °C
Density:0.89 g/cm3
Flash Pt:–21 °C
UN:2056
ADR:3,II
:GHS08
:GHS07
:GHS02
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356