


Trichloroacetic acid (TCA) 99-100.5% (dried basis), meets analytical specification of Ph. Eur., USP 21, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Boiling Pt:196 °C
Density:1.63 g/cm3
Flash Pt:113 °C
UN:1839
ADR:8,II
:GHS09
:GHS07
:GHS05
Specification Test Results
- Assay (calculated to the dried substance):99 - 100.5 %
- Loss on drying (on silica gel, 18h):Max. 0.5 %
- Sulfated ash:Max. 0.02 %
- Iron (Fe):Max. 0.002 %
- Chloride (Cl):Max. 0.005 %
- Sulfate (SO₄):Max. 0.05 %
- Appearance of the solution:complying