Order Entry
Germany
ContactUsLinkComponent
 
Glycine, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # HONC15527H-25KG
CAS Number:  
Glycine, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # HONC15527H-25KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Loss on drying (105°C, 2 h):
    Max. 0.2 %
  • Sulfated ash:
    Max. 0.1 %
  • pH (5 %, 20°C):
    5.9 - 6.4
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 0.005 %
  • Reaction against H₂SO₄:
    complying
  • Hydrolyzable substance:
    complying
  • Other ninhydr.-pos.S:
    Max. 0.5 %
  • Related substances (HPLC):
    complying
  • Identity (IR):
    complying
  • Appearance of the solution:
    complying
  • Appearance of the substance:
    complying
  • Residual solvents:
    complying

Specification Test Results

About this item

 :
 :75.07 g/mol
Density:1.160 g/cm3
CAS Number:56-40-6
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131