


N,N-Dimethylformamide, anhydrous 99.80%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Boiling Pt:153 °C
Melting Pt:–61 °C
Density:0.944
Flash Pt:57 °C
Storage Temperature:Ambient
UN:2265
ADR:3,III
:Packaged under argon.
EINECS:200-679-5
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless liquid
- Assay (GC):≥99.75%
- Water content (Karl Fischer Titration):≤50 ppm
- Residue on evaporation:≤5 ppm