Cut resistant gloves, Showa 542
Supplier: Showa
Certificates
About this item
These cut and abrasion resistant gloves made from HPPE protect hands during component assembly and hazardous applications
- Polyurethane coating over a HPPE reinforced liner
- Optimum flexibility and dexterity
- Open back of hand design reduces perspiration
- Seamless liner reduces irritation during long periods of wear
- Elasticated wrist cuff
These gloves were made with high performance Poly Ethylene (HPPE), coated palm, elasticated cuff, standard finish and ideally suited for aerospace, automotive, construction, manufacturing, ceramics, steel and metals. Typical application includes electrical appliance assembly, handling metal parts, precision mechanics, product protection, white goods handling and wire assembly.
Certifications: EN 388:2016 (4341B); Cat. II
Packaging: 10 pairs per polybag.
Specifications
- Catalog No:
- 112-5869
- 112-5868
- 112-5870
- 112-5867
- 112-5866
- Model:
- Showa 542
- Showa 542
- Showa 542
- Showa 542
- Showa 542
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Finish:
- PU coating
- PU coating
- PU coating
- PU coating
- PU coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4341B)
- EN 388:2016 (4341B)
- EN 388:2016 (4341B)
- EN 388:2016 (4341B)
- EN 388:2016 (4341B)