ContactUsLinkComponent
General purpose gloves, Showa 4400Y
General purpose gloves, Showa 4400Y
Supplier:  Showa
Certificates
General purpose gloves, Showa 4400Y
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5832
  • 112-5831
  • 112-5830
  • 112-5833
  • Model:
  • Showa 4400Y
  • Showa 4400Y
  • Showa 4400Y
  • Showa 4400Y
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Liner material:
  • Cotton jersey
  • Cotton jersey
  • Cotton jersey
  • Cotton jersey
  • Finish:
  • 3/4 nitrile coating
  • 3/4 nitrile coating
  • 3/4 nitrile coating
  • 3/4 nitrile coating
  • Cuff style:
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • EN norm:
  • EN 388:2016 (3111X)
  • EN 388:2016 (3111X)
  • EN 388:2016 (3111X)
  • EN 388:2016 (3111X)
Specifications