ContactUsLinkComponent
Impact resistant gloves, showa 377-IP
Impact resistant gloves, showa 377-IP
Supplier:  Showa
Certificates
Impact resistant gloves, showa 377-IP
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5815
  • 112-5814
  • 112-5816
  • 112-5813
  • Model:
  • 377-IP
  • 377-IP
  • 377-IP
  • 377-IP
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton/polyester
  • Cotton/polyester
  • Cotton/polyester
  • Cotton/polyester
  • Finish:
  • Fully coated (nitrile) + additional palm coating
  • Fully coated (nitrile) + additional palm coating
  • Fully coated (nitrile) + additional palm coating
  • Fully coated (nitrile) + additional palm coating
  • Cuff style:
  • Straight
  • Straight
  • Straight
  • Straight
  • EN norm:
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
Specifications