ContactUsLinkComponent
Safety spectacles, Contemporary 572 Diamond, small
Safety spectacles, Contemporary 572 Diamond, small
Catalog # 111-2459
Supplier:  UNIVET
undefined
Safety spectacles, Contemporary 572 Diamond, small
Catalog # 111-2459
Supplier:  UNIVET
Specifications
  • Type:
    Anti-UV, anti-scratch, anti-fog
  • Colour:
    Dark grey
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Contemporary series combines elegance and style in proetction, changing the concept of safety glasses.
  • Light and extremely tough
  • Integrated side shields
  • PA12 nylon frame with zero metallic components
  • Glossy varnish with shaded effect
  • Clear KN lenses, with anti-fog and anti-scratch ultra performing Vanguard PLUS treatment
Elegant and graceful for women, dynamic for men. Glasses can be employed for prolonged use as they do not create distorsions and allow a correct color perception, sensibly reducing visual fatigue.
Suitable for mechanical work in good visibility conditions.
Size: 52
Certifications: EN-166, EN-170.