Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
New Product
Certificates
About this item
These air valves can be used for analytical and preperative applications.
- PTFE filter for clean solvents
- Valve membrane blocks solvent vapours, opens at underpressure
- Thread size UNF 1/4" 28G
They include a luer lock adapter which is useful for many applications.