Order Entry
Germany
ContactUsLinkComponent
Hydrogen chloride solution 0.5 M in methanol, LiChropur™ for GC derivatisation, Supelco®
Flammable
Corrosive
Toxic
Health
Hydrogen chloride solution 0.5 M in methanol, LiChropur™ for GC derivatisation, Supelco®
Supplier:  Merck
Certificates
Flammable
Corrosive
Toxic
Health
Hydrogen chloride solution 0.5 M in methanol, LiChropur™ for GC derivatisation, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Appearance (Color):
    Colorless
  • Appearance (Form):
    Liquid
  • Titration (Method):
    Alkalimetric
  • Titration (Molarity):
    0.45 - 0.55 mol/l
  • Remarks on GC:
    Suitibility for GC derivatization complying
  • Density D20/4:
    0.800 - 0.811
  • Proton NMR Spectrum:
    Conforms to Structure
  • Recommended Retest Period:
    54 Months

Specification Test Results

Related Information

Frequently Bought Together