Order Entry
Germany
ContactUsLinkComponent
penta-Sodium triphosphate ≥98.0% (by titrimetric analysis), Purum p.a., Sigma-Aldrich®
penta-Sodium triphosphate ≥98.0% (by titrimetric analysis), Purum p.a., Sigma-Aldrich®
Catalog # SIAL72061-500G
Supplier:  Merck
CAS Number:  
penta-Sodium triphosphate ≥98.0% (by titrimetric analysis), Purum p.a., Sigma-Aldrich®
Catalog # SIAL72061-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless or White
  • Appearance (Form):
    Powder
  • Titration (Ion Exchange):
    57-59 % calc. based on P₂O₅
  • Titration:
    ≥ 98.0 %
  • Solubility (Color):
    Colorless
  • Solubility (Turbidity):
    Clear
  • Solubility (Method):
    0.1 g IN 10 ml Water
  • pH:
    9.0 - 10.0
  • Metal Trace Analysis (ICP):
    Corresponds to Requirements
  • Calcium (ICP):
    ≤ 200 mg/kg
  • Cadmium (ICP):
    ≤ 50 mg/kg
  • Cobalt (ICP):
    ≤ 50 mg/kg
  • Copper (ICP):
    ≤ 50 mg/kg
  • Iron (ICP):
    ≤ 50 mg/kg
  • Nickel (ICP):
    ≤ 50 mg/kg
  • Lead (ICP):
    ≤ 50 mg/kg
  • Zinc (ICP):
    ≤ 50 mg/kg
  • Total Sulfur As SO₄ (ICP):
    ≤ 250 mg/kg
  • Chloride (Cl):
    ≤ 1000 mg/k

Specification Test Results

About this item

 :
 :367.86 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:231-838-7
CAS Number:7758-29-4
Formula:Na₅O₁₀P₃
MDL Number:MFCD00003514