Order Entry
Germany
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Catalog # 1.06107.0003
Supplier:  Merck
CAS Number:  
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Catalog # 1.06107.0003
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic tube
  • Pk:
    3
  • Seq:
    0003

Specifications

Specification Test Results

  • Appearance:
    passes test

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:308080-99-1
 :1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613