Single-channel pipettes, mechanical, fixed / variable volume, Transferpette® S
Supplier: Brand
Certificates
About this item
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
- True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
- Colour code on volume adjustment wheel, shaft and volume-change protection
- Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
- Slim shaft for sampling from narrow vessels (centrifuge tubes)
- Universal tip cone
- Fully autoclavable at 121 °C
The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
Specifications
- Catalog No:
- BRNDDAKKS705870
- 613-6841
- BRNDDAKKS705878
- 613-6840
- 613-6837
- 613-6836
- 613-6839
- BRNDDAKKS705884
- 613-6838
- 613-6736
- 613-6833
- 613-6737
- 613-6832
- 613-6735
- 613-6835
- 613-6834
- 613-6738
- 613-6739
- BRNDDAKKS705873
- BRNDDAKKS705882
- BRNDDAKKS705862
- BRNDDAKKS705880
- BRNDDAKKS705874
- BRNDDAKKS705844
- 613-6734
- 613-6848
- 613-6847
- 613-6850
- 613-6849
- 613-6844
- 613-6843
- 613-6846
- 613-6845
- 613-6842
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Sustainable Packaging, Low Manufacturing Impact
- Low Manufacturing Impact, Sustainable Packaging
- Sustainable Packaging, Low Manufacturing Impact
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
Specifications
Related Information
Certificates
Search by Lot NumberBrand Mechanical, Transferpette® S
Product Literature
Brand Transferpette® S
Videos
Manual Transferpette® S
Manual
TDS Transferpette S 2020
Datasheet
Frequently Bought Together