Tetrabutylammonium methanesulphonate ≥97.0% (by titrimetric analysis), Supelco®
Catalog # 86877-10G
Supplier: Merck
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:337.56 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:65411-49-6
Formula:[CH₃CH₂CH₂CH₂]₄N(CH₃SO₃)
MDL Number:MFCD00043226