Methoxypolyethylene glycol 5,000 acetic acid N-succinimidyl ester ≥80%, Supelco®
Catalog # 85973-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Freezer
CAS Number:92451-01-9
MDL Number:MFCD01867746