Order Entry
Germany
ContactUsLinkComponent
Sodium bicarbonate : Sodium carbonate (100 mM : 100 mM) eluent concentrate for IC (20x) for Metrosep A Supp 10, Supelco®
Sodium bicarbonate : Sodium carbonate (100 mM : 100 mM) eluent concentrate for IC (20x) for Metrosep A Supp 10, Supelco®
Catalog # 75335-1L
Supplier:  Merck
CAS Number:  
Sodium bicarbonate : Sodium carbonate (100 mM : 100 mM) eluent concentrate for IC (20x) for Metrosep A Supp 10, Supelco®
Catalog # 75335-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Formula:Na₂CO₃/NaHCO₃