Order Entry
Germany
ContactUsLinkComponent
Titanium(IV) oxysulphate solution for hydrogen peroxide (H₂O₂) determination according to DIN 38 409, part 15 DEV-18, Supelco®
Titanium(IV) oxysulphate solution for hydrogen peroxide (H₂O₂) determination according to DIN 38 409, part 15 DEV-18, Supelco®
Catalog # 34244-1L
Supplier:  Merck
CAS Number:  
undefined
Titanium(IV) oxysulphate solution for hydrogen peroxide (H₂O₂) determination according to DIN 38 409, part 15 DEV-18, Supelco®
Catalog # 34244-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :159.93 g/mol (anhy)
 :M-Clarity™ quality level = MQ100
CAS Number:13825-74-6
Formula:TiOSO₄·aq
MDL Number:MFCD00149873