Orders LinkContactUsLinkComponent
Control units
Control units
Supplier:   2MAG
Control units
Supplier:  2MAG
Certificates
Control units
4 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 442-0769
  • 442-0768
  • 442-0770
  • TMAG90210_CH
  • TMAG90210_EU
  • TMAG90210_UK
  • 442-0772
  • 442-0771
  • 442-0773
  • TMAG90410_CH
  • TMAG90410_EU
  • TMAG90410_UK
  • Model:
  • MIXcontrol 20
  • MIXcontrol 20
  • MIXcontrol 20
  • MIXcontrol 20
  • MIXcontrol 20
  • MIXcontrol 20
  • MIXcontrol 40
  • MIXcontrol 40
  • MIXcontrol 40
  • MIXcontrol 40
  • MIXcontrol 40
  • MIXcontrol 40
  • Speed range:
  • 100 - 1600 min⁻¹
  • 100 - 1600 min⁻¹
  • 100 - 1600 min⁻¹
  • 100 - 1600 min⁻¹
  • 100 - 1600 min⁻¹
  • 100 - 1600 min⁻¹
  • 100 - 2000 min⁻¹
  • 100 - 2000 min⁻¹
  • 100 - 2000 min⁻¹
  • 100 - 2000 min⁻¹
  • 100 - 2000 min⁻¹
  • 100 - 2000 min⁻¹
  • Power input:
  • 20 W
  • 20 W
  • 20 W
  • 20 W
  • 20 W
  • 20 W
  • 40 W
  • 40 W
  • 40 W
  • 40 W
  • 40 W
  • 40 W
  • Weight:
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • 1,9 kg
  • W×D×H:
  • 200×155×38 mm
  • 200×155×38 mm
  • 200×155×38 mm
  • 200×155×38 mm
  • 200×155×38 mm
  • 200×155×38 mm
  • 200×155×48 mm
  • 200×155×48 mm
  • 200×155×48 mm
  • 200×155×48 mm
  • 200×155×48 mm
  • 200×155×48 mm
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 442-0769
    MIXcontrol 20, CH-plug
    MIXcontrol 20, CH-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    442-0769
    Model
    MIXcontrol 20
    Speed range
    100 - 1600 min⁻¹
    Power input
    20 W
    Weight
    1,9 kg
    W×D×H
    200×155×38 mm
  • 442-0768
    MIXcontrol 20, EU-plug
    MIXcontrol 20, EU-plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 748,00 €
    748,00 €
    1 * 1 ST
       748,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    442-0768
    Model
    MIXcontrol 20
    Speed range
    100 - 1600 min⁻¹
    Power input
    20 W
    Weight
    1,9 kg
    W×D×H
    200×155×38 mm
  • 442-0770
    MIXcontrol 20, UK-plug
    MIXcontrol 20, UK-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    442-0770
    Model
    MIXcontrol 20
    Speed range
    100 - 1600 min⁻¹
    Power input
    20 W
    Weight
    1,9 kg
    W×D×H
    200×155×38 mm
  • TMAG90210_CH
    MIXcontrol 20, with RS232 interface, CH-plug
    MIXcontrol 20, with RS232 interface, CH-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    TMAG90210_CH
    Model
    MIXcontrol 20
    Speed range
    100 - 1600 min⁻¹
    Power input
    20 W
    Weight
    1,9 kg
    W×D×H
    200×155×38 mm
  • TMAG90210_EU
    MIXcontrol 20, with RS232 interface, EU-plug
    MIXcontrol 20, with RS232 interface, EU-plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 906,00 €
    906,00 €
    1 * 1 ST
       906,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    TMAG90210_EU
    Model
    MIXcontrol 20
    Speed range
    100 - 1600 min⁻¹
    Power input
    20 W
    Weight
    1,9 kg
    W×D×H
    200×155×38 mm
  • TMAG90210_UK
    MIXcontrol 20, with RS232 interface, UK-plug
    MIXcontrol 20, with RS232 interface, UK-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    TMAG90210_UK
    Model
    MIXcontrol 20
    Speed range
    100 - 1600 min⁻¹
    Power input
    20 W
    Weight
    1,9 kg
    W×D×H
    200×155×38 mm
  • 442-0772
    MIXcontrol 40, CH-plug
    MIXcontrol 40, CH-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    442-0772
    Model
    MIXcontrol 40
    Speed range
    100 - 2000 min⁻¹
    Power input
    40 W
    Weight
    1,9 kg
    W×D×H
    200×155×48 mm
  • 442-0771
    MIXcontrol 40, EU-plug
    MIXcontrol 40, EU-plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    442-0771
    Model
    MIXcontrol 40
    Speed range
    100 - 2000 min⁻¹
    Power input
    40 W
    Weight
    1,9 kg
    W×D×H
    200×155×48 mm
  • 442-0773
    MIXcontrol 40, UK-plug
    MIXcontrol 40, UK-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    442-0773
    Model
    MIXcontrol 40
    Speed range
    100 - 2000 min⁻¹
    Power input
    40 W
    Weight
    1,9 kg
    W×D×H
    200×155×48 mm
  • TMAG90410_CH
    MIXcontrol 40, with RS232 interface, CH-plug
    MIXcontrol 40, with RS232 interface, CH-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    TMAG90410_CH
    Model
    MIXcontrol 40
    Speed range
    100 - 2000 min⁻¹
    Power input
    40 W
    Weight
    1,9 kg
    W×D×H
    200×155×48 mm
  • TMAG90410_EU
    MIXcontrol 40, with RS232 interface, EU-plug
    MIXcontrol 40, with RS232 interface, EU-plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 200,00 €
    1 200,00 €
    1 * 1 ST
       1 200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    TMAG90410_EU
    Model
    MIXcontrol 40
    Speed range
    100 - 2000 min⁻¹
    Power input
    40 W
    Weight
    1,9 kg
    W×D×H
    200×155×48 mm
  • TMAG90410_UK
    MIXcontrol 40, with RS232 interface, UK-plug
    MIXcontrol 40, with RS232 interface, UK-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    TMAG90410_UK
    Model
    MIXcontrol 40
    Speed range
    100 - 2000 min⁻¹
    Power input
    40 W
    Weight
    1,9 kg
    W×D×H
    200×155×48 mm