Order Entry
Germany
ContactUsLinkComponent
 
Peqlab peqGOLD, DNA Isolation, Bacterial DNA Mini Kit
 
undefined
Peqlab peqGOLD, DNA Isolation, Bacterial DNA Mini Kit
 

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Sample size:
    3 ml log phase culture or 1×10⁹ cells
  • Description:
    Bacterial DNA Mini Kit