Order Entry
Germany
ContactUsLinkComponent
Capping Reagent 80/10/10 THF/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent 80/10/10 THF/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR640-4X2.5L
CAS Number:  
Capping Reagent 80/10/10 THF/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR640-4X2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 L
  • Seq:
    5114
  • Pk Info:
    4x 2.5 L

Specifications

About this item

 :
Boiling Pt:11.8 %(V) (main component)
Melting Pt:–108.5 °C (pure substance)
Density:0.888 g/cm³ (20 °C) (main component)
Flash Pt:–15 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2924
ADR:3,II
 :GHS02
 :GHS08
 :GHS05
 :GHS07
CAS Number:109-99-9